Learn More
Biomatik Corporation Mouse Fatty Acid Binding Protein 7, Brain (FABP7) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU09054
Encompass_Preferred
Mouse Fatty Acid Binding Protein 7, Brain (FABP7) ELISA Kit for detecting FABP7 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 62.5-4,000pg/ml. Sensitivity: 26.3pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P51880. Gene ID: 12140. Target Synonyms: B-FABP; BLBP; FABPB; MRG; Brain Lipid Binding Protein; Mammary-derived growth inhibitor related. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4333