Learn More
Biomatik Corporation Human Colony Stimulating Factor Receptor, Macrophage (MCSFR) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU09066
Encompass_Preferred
The Human Colony Stimulating Factor Receptor, Macrophage (MCSFR) ELISA Kit allows for quantitative determination of Colony Stimulating Factor Receptor, Macrophage in Tissue homogenates, Cell lysates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.312-20 ng/mL. Sensitivity: 0.128 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P07333. Cross Activity: No significant cross-reactivity or interference between Colony Stimulating Factor Receptor, Macrophage and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-4336