Learn More
Biomatik Corporation Human Large Multifunctional Peptidase 7 (LMP7) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU09068
Encompass_Preferred
The Human Large Multifunctional Peptidase 7 (LMP7) ELISA Kit allows for quantitative determination of Large Multifunctional Peptidase 7 in Tissue homogenates, Cell lysates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.312-20 ng/mL. Sensitivity: 0.118 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P28062. Cross Activity: No significant cross-reactivity or interference between Large Multifunctional Peptidase 7 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-4337