Learn More
Biomatik Corporation Mouse Surfactant Associated Protein B (SPB) ELISA Kit, 96 tests
Catalog No. 501504338
Encompass_Preferred
Mouse Surfactant Protein B (SP-B) ELISA Kit for detecting SP-B in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.78-50ng/ml. Sensitivity: 0.32ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P50405. Gene ID: 20388. Target Synonyms: SFTPB; PSPB; SFTB3; SFTP3; SPB; Surfactant Associated Protein B; Pulmonary Surfactant Protein B; 18 kDa pulmonary-surfactant protein; 6 kDa protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4338 Supplier Biomatik Corporation Supplier No. EKU09070
Mouse Surfactant Protein B (SP-B) ELISA Kit for detecting SP-B in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.78-50ng/ml. Sensitivity: 0.32ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P50405. Gene ID: 20388. Target Synonyms: SFTPB; PSPB; SFTB3; SFTP3; SPB; Surfactant Associated Protein B; Pulmonary Surfactant Protein B; 18 kDa pulmonary-surfactant protein; 6 kDa protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.