Learn More
Biomatik Corporation Mouse Tryptophanyl tRNA Synthetase (WARS) ELISA Kit, 96 tests
Catalog No. 501504340
Encompass_Preferred
Mouse Tryptophanyl tRNA Synthetase (WARS) ELISA Kit for detecting WARS in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.064ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P32921. Gene ID: 22375. Target Synonyms: GAMMA-2; IFI53; IFP53; WRS; TrpRS; hWRS; Tryptophan tRNA Ligase 1,Cytoplasmic; Interferon-induced protein 53; Tryptophanyl-tRNA synthetase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4340 Supplier Biomatik Corporation Supplier No. EKU09081
Mouse Tryptophanyl tRNA Synthetase (WARS) ELISA Kit for detecting WARS in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.064ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P32921. Gene ID: 22375. Target Synonyms: GAMMA-2; IFI53; IFP53; WRS; TrpRS; hWRS; Tryptophan tRNA Ligase 1,Cytoplasmic; Interferon-induced protein 53; Tryptophanyl-tRNA synthetase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.