Learn More
Biomatik Corporation Human Keratin 17 (KRT17) ELISA Kit, 96 tests
Catalog No. 501504341
Encompass_Preferred
The Human Keratin 17 (KRT17) ELISA Kit allows for quantitative determination of Keratin 17 in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 78-5000 pg/mL. Sensitivity: 30 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q04695. Cross Activity: No significant cross-reactivity or interference between Keratin 17 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-4341 Supplier Biomatik Corporation Supplier No. EKU09088
The Human Keratin 17 (KRT17) ELISA Kit allows for quantitative determination of Keratin 17 in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 78-5000 pg/mL. Sensitivity: 30 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q04695. Cross Activity: No significant cross-reactivity or interference between Keratin 17 and analogues was observed. Restriction: For Research Use Only.