Learn More
Biomatik Corporation Multi-species Noggin (NOG) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU09106
Encompass_Preferred
Human/Mouse/Rat Noggin (NOG) ELISA Kit for detecting NOG in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: 1.56-100ng/ml. Sensitivity: 0.62ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q13253. Gene ID: 9241. Target Synonyms: SYM1; SYNS1; Synostoses (Multiple) Syndrome 1; Symphalangism 1 (Proximal). Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4345