Learn More
Biomatik Corporation Mouse Protein Phosphatase 1, Regulatory Subunit 15A (PPP1R15A) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU09109
Encompass_Preferred
Mouse Protein Phosphatase 1, Regulatory Subunit 15A (PPP1R15A) ELISA Kit for detecting PPP1R15A in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.056ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P17564. Gene ID: 17872. Target Synonyms: PPP1-R15A; PPP1R15-A; PPP1-R15-A; GADD34; Growth Arrest And DNA-Damage-Inducible 34; Myeloid differentiation primary response protein MyD116 homolog. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4347