Learn More
Biomatik Corporation Human Urocortin 2 (UCN2) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU09129
Encompass_Preferred
Human Urocortin 2 (UCN2) ELISA Kit for detecting UCN2 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.78-50ng/ml. Sensitivity: 0.29ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q96RP3. Gene ID: 90226. Target Synonyms: URP; SRP; UCN-II; UCNII; Stresscopin-Related Peptide; Urocortin-related peptide. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4352