Learn More
Biomatik Corporation Rat Rhodopsin (RHO) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU09138
Encompass_Preferred
Rat Rhodopsin (RHO) ELISA Kit for detecting RHO in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.057ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P51489. Gene ID: 24717. Target Synonyms: OPN2; RP4; Opsin 2; Rod Pigment; Retinitis Pigmentosa 4; Visual Purple. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4354