Learn More
Biomatik Corporation Human Thioredoxin Reductase 2 (TrxR2) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU09162
Encompass_Preferred
Human Thioredoxin Reductase 2 (TrxR2) ELISA Kit for detecting TrxR2 in serum, plasma, tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.113ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9NNW7. Gene ID: 10587. Target Synonyms: TR; TXNRD2; TR3; SelZ; Thioredoxin Reductase Beta; Selenoprotein Z. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4358