Learn More
Biomatik Corporation Mouse Dickkopf Related Protein 3 (DKK3) ELISA Kit, 96 tests
Catalog No. 501504359
Encompass_Preferred
Mouse Dickkopf Related Protein 3 (DKK3) ELISA Kit for detecting DKK3 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.065ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9QUN9. Gene ID: 50781. Target Synonyms: REIC. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4359 Supplier Biomatik Corporation Supplier No. EKU09164
Mouse Dickkopf Related Protein 3 (DKK3) ELISA Kit for detecting DKK3 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.065ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9QUN9. Gene ID: 50781. Target Synonyms: REIC. Restrictions: For Research Use Only. Not for use in diagnostic procedures.