Learn More
Biomatik Corporation Human Aryl Hydrocarbon Receptor Nuclear Translocator Like Protein (ARNTL) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU09165
Encompass_Preferred
The Human Aryl Hydrocarbon Receptor Nuclear Translocator Like Protein (ARNTL) ELISA Kit allows for quantitative determination of Aryl Hydrocarbon Receptor Nuclear Translocator Like Protein in Tissue homogenates, Cell lysates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.312-20 ng/mL. Sensitivity: 0.112 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: O00327. Cross Activity: No significant cross-reactivity or interference between Aryl Hydrocarbon Receptor Nuclear Translocator Like Protein and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-4360