Learn More
Biomatik Corporation Pig N-Terminal Pro-Brain Natriuretic Peptide (NT-ProBNP) ELISA Kit
Supplier:  Biomatik Corporation EKU10175
Encompass_Preferred
Pig N-Terminal Pro-Brain Natriuretic Peptide (NT-ProBNP) ELISA Kit for detecting NT-ProBNP in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Pig (Sus scrofa; Porcine). Detection Range: 61.7-5,000pg/ml. Sensitivity: 28.8pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: P07634. Gene ID: 396844. Target Synonyms: NT-Pro-BNP; ; N-BNP. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6669