Learn More
Biomatik Corporation Rat Glucagon Like Peptide 1 Receptor (GLP1R) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU09167
Encompass_Preferred
Rat Glucagon Like Peptide 1 Receptor (GLP1R) ELISA Kit for detecting GLP1R in tissue homogenates, cell lysates other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.055ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P32301. Gene ID: 25051. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4361