Learn More
Biomatik Corporation Human Signal Transducer And Activator Of Transcription 1 (STAT1) ELISA Kit
Supplier:  Biomatik Corporation EKU10223
Encompass_Preferred
Human Signal Transducer And Activator Of Transcription 1 (STAT1) ELISA Kit for detecting STAT1 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.057ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P42224. Gene ID: 6772. Target Synonyms: ISGF-3; STAT91; Transcription Factor ISGF-3 Components p91/p84. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6678