Learn More
Biomatik Corporation Rat Urate Transporter 1 (URAT1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU09195
Encompass_Preferred
Rat Urate Transporter 1 (URAT1) ELISA Kit for detecting URAT1 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.625-40ng/ml. Sensitivity: 0.285ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q3ZAV1. Gene ID: 365398. Target Synonyms: SLC22A12; OAT4L; RST; Solute Carrier Family 22 Member 12,Organic Anion/Urate Transporter; Organic anion transporter 4-like; Urate anion exchanger 1; Renal-specific transporter. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4365