Learn More
Biomatik Corporation Human Uroplakin 2 (UPK2) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU09219
Encompass_Preferred
Human Uroplakin 2 (UPK2) ELISA Kit for detecting UPK2 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.055ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O00526. Gene ID: 7379. Target Synonyms: UPII; Uroplakin II. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4369