Learn More
Biomatik Corporation Human Sestrin 3 (SESN3) ELISA Kit, 96 tests
Catalog No. 501504371
Encompass_Preferred
Human Sestrin 3 (SESN3) ELISA Kit for detecting SESN3 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.119ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P58005. Gene ID: 143686. Target Synonyms: SEST3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4371 Supplier Biomatik Corporation Supplier No. EKU09223
Human Sestrin 3 (SESN3) ELISA Kit for detecting SESN3 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.119ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P58005. Gene ID: 143686. Target Synonyms: SEST3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.