Learn More
Biomatik Corporation Human 7-Dehydrocholesterol (7-DHC) ELISA Kit
Supplier:  Biomatik Corporation EKU10601
Encompass_Preferred
Human 7-Dehydrocholesterol (7-DHC) ELISA Kit for detecting 7-DHC in serum, plasma, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 2.47-200ng/ml. Sensitivity: 0.91ng/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6714