Learn More
Biomatik Corporation Human Vitamin K Epoxide Reductase Complex Subunit 1 (VKORC1) ELISA Kit, 96 tests
Catalog No. 501504373
Encompass_Preferred
Human Vitamin K Epoxide Reductase Complex Subunit 1 (VKORC1) ELISA Kit for detecting VKORC1 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 1.56-100ng/ml. Sensitivity: 0.60ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9BQB6. Gene ID: 79001. Target Synonyms: VKCFD2; VKOR; Vitamin K Dependent Clotting Factors Deficiency 2; Vitamin K1 2,3-epoxide reductase subunit 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4373 Supplier Biomatik Corporation Supplier No. EKU09239
Human Vitamin K Epoxide Reductase Complex Subunit 1 (VKORC1) ELISA Kit for detecting VKORC1 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 1.56-100ng/ml. Sensitivity: 0.60ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9BQB6. Gene ID: 79001. Target Synonyms: VKCFD2; VKOR; Vitamin K Dependent Clotting Factors Deficiency 2; Vitamin K1 2,3-epoxide reductase subunit 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.