Learn More
Biomatik Corporation Human Vasohibin 1 (VASH1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU09240
Encompass_Preferred
Human Vasohibin 1 (VASH1) ELISA Kit for detecting VASH1 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.625-40ng/ml. Sensitivity: 0.224ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q7L8A9. Gene ID: 22846. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4374