Learn More
Biomatik Corporation Human S100 Calcium Binding Protein B (S100B) Anti-antigen ELISA Kit
Supplier:  Biomatik Corporation EKU10648
Encompass_Preferred
Human S100 Calcium Binding Protein B (S100B) Anti-antigen ELISA Kit for detecting S100B in serum, plasma, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 3.12-200ng/ml. Sensitivity: 1.25ng/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h, 30min. Accession Number: P04271. Gene ID: 6285. Target Synonyms: S100-B; NEF; S100Beta; S-100 protein subunit beta. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6730