Learn More
Biomatik Corporation Human Ras Association Domain Containing Protein 1 (RASSF1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU09283
Encompass_Preferred
The Human Ras Association Domain Containing Protein 1 (RASSF1) ELISA Kit allows for quantitative determination of Ras Association Domain Containing Protein 1 in Tissue homogenates, Cell lysates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.156-10 ng/mL. Sensitivity: 0.059 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q9NS23. Cross Activity: No significant cross-reactivity or interference between Ras Association Domain Containing Protein 1 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-4380