Learn More
Biomatik Corporation Human Cytochrome P450 4F2 (CYP4F2) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU09309
Encompass_Preferred
The Human Cytochrome P450 4F2 (CYP4F2) ELISA Kit allows for quantitative determination of Cytochrome P450 4F2 in Tissue homogenates, Cell lysates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 1.56-100 ng/mL. Sensitivity: 0.55 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P78329. Cross Activity: No significant cross-reactivity or interference between Cytochrome P450 4F2 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-4383