Learn More
Biomatik Corporation Human Protein Kinase, cAMP Dependent Catalytic Alpha (PRKACa) ELISA Kit, 96 tests
Catalog No. 501504384
Encompass_Preferred
The Human Protein Kinase, cAMP Dependent Catalytic Alpha (PRKACa) ELISA Kit allows for quantitative determination of Protein Kinase, cAMP Dependent Catalytic Alpha in Tissue homogenates, Cell lysates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.156-10 ng/mL. Sensitivity: 0.058 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P17612. Cross Activity: No significant cross-reactivity or interference between Protein Kinase, cAMP Dependent Catalytic Alpha and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-4384 Supplier Biomatik Corporation Supplier No. EKU09325
The Human Protein Kinase, cAMP Dependent Catalytic Alpha (PRKACa) ELISA Kit allows for quantitative determination of Protein Kinase, cAMP Dependent Catalytic Alpha in Tissue homogenates, Cell lysates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.156-10 ng/mL. Sensitivity: 0.058 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P17612. Cross Activity: No significant cross-reactivity or interference between Protein Kinase, cAMP Dependent Catalytic Alpha and analogues was observed. Restriction: For Research Use Only.