Learn More
Biomatik Corporation Human Secreted Ly6/uPAR Related Protein 1 (SLURP1) ELISA Kit, 96 tests
Catalog No. 501504387
Encompass_Preferred
Human Secreted Ly6/uPAR Related Protein 1 (SLURP1) ELISA Kit for detecting SLURP1 in serum, plasma, urine, saliva, tears, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.058ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P55000. Gene ID: 57152. Target Synonyms: ARS; ANUP; ArsB; LY6LS; MDM; Lymphocyte Antigen 6-Like Secreted; ARS Component B; Anti-neoplastic urinary protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4387 Supplier Biomatik Corporation Supplier No. EKU09331
Human Secreted Ly6/uPAR Related Protein 1 (SLURP1) ELISA Kit for detecting SLURP1 in serum, plasma, urine, saliva, tears, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.058ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P55000. Gene ID: 57152. Target Synonyms: ARS; ANUP; ArsB; LY6LS; MDM; Lymphocyte Antigen 6-Like Secreted; ARS Component B; Anti-neoplastic urinary protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.