Learn More
Biomatik Corporation Human Specificity Protein 1 (Sp1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU09332
Encompass_Preferred
Human Specificity Protein 1 (Sp1) ELISA Kit for detecting Sp1 in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.064ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P08047. Gene ID: 6667. Target Synonyms: TSFP1; Sp1 Transcription Factor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4388