Learn More
Biomatik Corporation Human Testican 1 (TIC1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU09333
Encompass_Preferred
Human Testican 1 (TIC1) ELISA Kit for detecting TIC1 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.061ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q08629. Gene ID: 6695. Target Synonyms: SPOCK1; SPOCK; TESTICAN; TIC1; Sparc/Osteonectin,Swcv And Kazal-Like Domains Proteoglycan 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4389