Learn More
Biomatik Corporation Human DnaJ/HSP40 Homolog Subfamily B, Member 9 (DNAJB9) ELISA Kit
Supplier:  Biomatik Corporation EKU10797
Encompass_Preferred
Human DnaJ/HSP40 Homolog Subfamily B, Member 9 (DNAJB9) ELISA Kit for detecting DNAJB9 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.114ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9UBS3. Gene ID: 4189. Target Synonyms: ERdj4; MDG1; Endoplasmic reticulum DNA J domain-containing protein 4; Microvascular endothelial differentiation gene 1 protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6768