Learn More
Biomatik Corporation Mouse Wingless Type MMTV Integration Site Family, Member 16 (WNT16) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU09341
Encompass_Preferred
Mouse Wingless Type MMTV Integration Site Family, Member 16 (WNT16) ELISA Kit for detecting WNT16 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 78-5,000pg/ml. Sensitivity: 30pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9QYS1. Gene ID: 93735. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4392