Learn More
Biomatik Corporation Multi-species Wingless Type MMTV Integration Site Family, Member 5A (WNT5A) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU09343
Encompass_Preferred
Human/Mouse/Rat Wingless Type MMTV Integration Site Family, Member 5A (WNT5A) ELISA Kit for detecting WNT5A in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.059ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P41221. Gene ID: 7474. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4394