Learn More
Biomatik Corporation Human H2A Histone Family, Member X (H2AFX) ELISA Kit, 96 tests
Catalog No. 501504395
Encompass_Preferred
The Human H2A Histone Family, Member X (H2AFX) ELISA Kit allows for quantitative determination of H2A Histone Family, Member X in Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.156-10 ng/mL. Sensitivity: 0.059 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P16104. Cross Activity: No significant cross-reactivity or interference between H2A Histone Family, Member X and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-4395 Supplier Biomatik Corporation Supplier No. EKU09348
The Human H2A Histone Family, Member X (H2AFX) ELISA Kit allows for quantitative determination of H2A Histone Family, Member X in Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.156-10 ng/mL. Sensitivity: 0.059 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P16104. Cross Activity: No significant cross-reactivity or interference between H2A Histone Family, Member X and analogues was observed. Restriction: For Research Use Only.