Learn More
Biomatik Corporation Mouse Vanin 1 (VNN1) ELISA Kit, 96 tests
Catalog No. 501504402
Encompass_Preferred
Mouse Vanin 1 (VNN1) ELISA Kit for detecting VNN1 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.060ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9Z0K8. Gene ID: 22361. Target Synonyms: Pantetheinase; Pantetheine hydrolase; Tiff66; Vascular non-inflammatory molecule 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4402 Supplier Biomatik Corporation Supplier No. EKU09400
Mouse Vanin 1 (VNN1) ELISA Kit for detecting VNN1 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.060ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9Z0K8. Gene ID: 22361. Target Synonyms: Pantetheinase; Pantetheine hydrolase; Tiff66; Vascular non-inflammatory molecule 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.