Learn More
Biomatik Corporation Cattle Prolactin (PRL) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU09419
Encompass_Preferred
The Cattle Prolactin (PRL) ELISA Kit allows for quantitative determination of Prolactin in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Bos taurus; Bovine (Cattle). Detection Range: 3.12-200 ng/mL. Sensitivity: 1.41 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P01239. Cross Activity: No significant cross-reactivity or interference between Prolactin and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-4405