Learn More
Biomatik Corporation Human Bone Morphogenetic Protein 4 (BMP4) ELISA Kit (CLIA), 96 tests
Supplier:  Biomatik Corporation EKU09429
Encompass_Preferred
The Human Bone Morphogenetic Protein 4 (BMP4) ELISA Kit (CLIA) allows for quantitative determination of Bone Morphogenetic Protein 4 in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 2.74-2,000 pg/mL. Sensitivity: 1.15 pg/mL. Format: 96-Well Plate. Assay Type: CLIA, Double-antibody Sandwich. Detection Method: Chemiluminescent. Assay Time: 2 hours, 40minutes. UniProt ID: P12644. Cross Activity: No significant cross-reactivity or interference between Bone Morphogenetic Protein 4 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-4406