Learn More
Biomatik Corporation Human Matrix Metalloproteinase 3 (MMP3) ELISA Kit (CLIA), 96 tests
Supplier:  Biomatik Corporation EKU09447
Encompass_Preferred
The Human Matrix Metalloproteinase 3 (MMP3) ELISA Kit (CLIA) allows for quantitative determination of Matrix Metalloproteinase 3 in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 41.2-30,000 pg/mL. Sensitivity: 17.4 pg/mL. Format: 96-Well Plate. Assay Type: CLIA, Double-antibody Sandwich. Detection Method: Chemiluminescent. Assay Time: 2 hours, 40minutes. UniProt ID: P08254. Cross Activity: No significant cross-reactivity or interference between Matrix Metalloproteinase 3 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-4408