Learn More
Biomatik Corporation Human High Mobility Group Protein 1 (HMGB1) Instant ELISA Kit
Supplier:  Biomatik Corporation EKU11001
Encompass_Preferred
Human High Mobility Group Protein 1 (HMGB1) Instant ELISA Kit for detecting HMGB1 in serum, plasma, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 62.5-4,000pg/ml. Sensitivity: 22.5pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 1h, 10min. Accession Number: P09429. Gene ID: 3146. Target Synonyms: HMG1; HMG3; SBP1; Sulfoglucuronyl Carbohydrate Binding Protein; Amphoterin; High Mobility Group Box 1 Protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6831