Learn More
Biomatik Corporation Human Growth Hormone 2 (GH2) ELISA Kit
Supplier:  Biomatik Corporation EKU04613
Encompass_Preferred
Human Growth Hormone 2 (GH2) ELISA Kit for detecting GH2 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 78-5,000pg/ml. Sensitivity: 33pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P01242. Gene ID: 2689. Target Synonyms: GH-V; GHL; GHV; hGH-V; Growth Hormone Variant; Placenta-Specific Growth Hormone; Placental Specific Growth Hormone. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6058