Learn More
Biomatik Corporation Human Cat Eye Syndrome Chromosome Region, Candidate 1 (CECR1) ELISA Kit (CLIA), 96 tests
Supplier:  Biomatik Corporation EKU09496
Encompass_Preferred
The Human Cat Eye Syndrome Chromosome Region, Candidate 1 (CECR1) ELISA Kit (CLIA) allows for quantitative determination of Cat Eye Syndrome Chromosome Region, Candidate 1 in Serum, Plasma and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 27.4-20,000 pg/mL. Sensitivity: 9.5 pg/mL. Format: 96-Well Plate. Assay Type: CLIA, Double-antibody Sandwich. Detection Method: Chemiluminescent. Assay Time: 2 hours, 40minutes. UniProt ID: Q9NZK5. Cross Activity: No significant cross-reactivity or interference between Cat Eye Syndrome Chromosome Region, Candidate 1 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-4412