Learn More
Biomatik Corporation Dog Matrix Metalloproteinase 9 (MMP9) ELISA Kit
Supplier:  Biomatik Corporation EKU05879
Encompass_Preferred
Dog Matrix Metalloproteinase 9 (MMP9) ELISA Kit for detecting MMP9 in serum, plasma, other biological fluids. Species Reactivity: Dog (Canis familiaris; Canine). Detection Range: 3.12-200ng/ml. Sensitivity: 1.22ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O18733. Target Synonyms: GELB; Gelatinase B; CLG4B; CLG4-B; 92 KDa Gelatinase; 92kDa Type IV Collagenase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6217