Learn More
Biomatik Corporation Mouse Cathepsin D (CTSD) ELISA Kit (CLIA), 96 tests
Supplier:  Biomatik Corporation EKU09500
Encompass_Preferred
Mouse Cathepsin D (CTSD) CLIA Kit for detecting CTSD in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 2.74-2,000pg/ml. Sensitivity: 0.98pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P18242. Gene ID: 13033. Target Synonyms: CPSD; CLN10; Lysosomal Aspartyl Protease; Ceroid-Lipofuscinosis,Neuronal 10. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4414