Learn More
Biomatik Corporation Rat Amphiregulin (AREG) ELISA Kit (CLIA), 96 tests
Supplier:  Biomatik Corporation EKU09519
Encompass_Preferred
Rat Amphiregulin (AREG) CLIA Kit for detecting AREG in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 1.37-1000pg/ml. Sensitivity: 0.58pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P24338. Gene ID: 29183. Target Synonyms: AR; CRDGF; SDGF; Colorectum Cell-Derived Growth Factor; Schwannoma-Derived Growth Factor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4416