Learn More
Biomatik Corporation Rat Connective Tissue Growth Factor (CTGF) ELISA Kit (CLIA), 96 tests
Catalog No. 501504418
Encompass_Preferred
Rat Connective Tissue Growth Factor (CTGF) CLIA Kit for detecting CTGF in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 5.49-4000pg/ml. Sensitivity: 1.99pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: Q9R1E9. Gene ID: 64032. Target Synonyms: CCN2; HCS24; IGFBP8; NOV2; Insulin-Like Growth Factor-Binding Protein 8; Hypertrophic Chondrocyte-Specific Protein 24; CCN Family Member 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4418 Supplier Biomatik Corporation Supplier No. EKU09524
Rat Connective Tissue Growth Factor (CTGF) CLIA Kit for detecting CTGF in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 5.49-4000pg/ml. Sensitivity: 1.99pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: Q9R1E9. Gene ID: 64032. Target Synonyms: CCN2; HCS24; IGFBP8; NOV2; Insulin-Like Growth Factor-Binding Protein 8; Hypertrophic Chondrocyte-Specific Protein 24; CCN Family Member 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.