Learn More
Biomatik Corporation Human Fibroblast Growth Factor 21 (FGF21) ELISA Kit (CLIA), 96 tests
Supplier:  Biomatik Corporation EKU09535
Encompass_Preferred
The Human Fibroblast Growth Factor 21 (FGF21) ELISA Kit (CLIA) allows for quantitative determination of Fibroblast Growth Factor 21 in Serum, Plasma and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 2.74-2,000 pg/mL. Sensitivity: 1.15 pg/mL. Format: 96-Well Plate. Assay Type: CLIA, Double-antibody Sandwich. Detection Method: Chemiluminescent. Assay Time: 2 hours, 40minutes. UniProt ID: Q9NSA1. Cross Activity: No significant cross-reactivity or interference between Fibroblast Growth Factor 21 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-4419