Learn More
Biomatik Corporation Mouse Matrix Metalloproteinase 1 (MMP1) ELISA Kit (CLIA), 96 tests
Supplier:  Biomatik Corporation EKU09559
Encompass_Preferred
Mouse Matrix Metalloproteinase 1 (MMP1) CLIA Kit for detecting MMP1 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 27.4-20,000pg/ml. Sensitivity: 9.7pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: Q9EPL5. Gene ID: 83995. Target Synonyms: CLGN; CLG1; Collagenase; Interstitial Collagenase; Vertebrate Collagenase; Fibroblast Collagenase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4421