Learn More
Biomatik Corporation Human Matrix Metalloproteinase 13 (MMP13) ELISA Kit (CLIA), 96 tests
Supplier:  Biomatik Corporation EKU09574
Encompass_Preferred
The Human Matrix Metalloproteinase 13 (MMP13) ELISA Kit (CLIA) allows for quantitative determination of Matrix Metalloproteinase 13 in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 54.9-40000 pg/mL. Sensitivity: 19.3 pg/mL. Format: 96-Well Plate. Assay Type: CLIA, Double-antibody Sandwich. Detection Method: Chemiluminescent. Assay Time: 2 hours, 40minutes. UniProt ID: P45452. Cross Activity: No significant cross-reactivity or interference between Matrix Metalloproteinase 13 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-4423