Learn More
Biomatik Corporation Human Signal Regulatory Protein Alpha (SIRPa) ELISA Kit
Supplier:  Biomatik Corporation EKU07332
Encompass_Preferred
Sample Type: Tissue homogenates, cell lysates and other biological fluids. Species Reactivity: Human. Detection Range: 0.156-10ng/mL. Sensitivity: 0.059ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: P78324. Target Synonyms: CD172a; SIRP-A; P84; BIT; MFR; MYD1; PTPNS1; SHPS1; SIRP; Tyrosine-Protein Phosphatase Non-Receptor Type Substrate 1; Macrophage fusion receptor. Restriction: For Research Use Only.
Catalog No. 50-228-6382