Learn More
Biomatik Corporation Mouse Matrix Metalloproteinase 13 (MMP13) ELISA Kit (CLIA), 96 tests
Supplier:  Biomatik Corporation EKU09575
Encompass_Preferred
Mouse Matrix Metalloproteinase 13 (MMP13) CLIA Kit for detecting MMP13 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 27.4-20,000pg/ml. Sensitivity: 11.1pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P33435. Gene ID: 17386. Target Synonyms: CLG3; Collagenase 3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4424