Learn More
Biomatik Corporation Human Sirtuin 7 (SIRT7) ELISA Kit
Supplier:  Biomatik Corporation EKU07359
Encompass_Preferred
Sample Type: tissue homogenates, cell lysates, cell culture supernates and other biological fluids. Species Reactivity: Human. Detection Range: 0.78-50ng/mL. Sensitivity: 0.27ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: Q9NRC8. Target Synonyms: SIR2L7; NAD-Dependent Deacetylase Sirtuin-7; Silent Mating Type Information Regulation 2 Homolog 7; Regulatory protein SIR2 homolog 7; SIR2-like protein 7. Restriction: For Research Use Only.
Catalog No. 50-228-6386